Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGCAGTCAGCGTCACCGTGGGG[A/G]GCAAGCAGTACCTCCTAGGACTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605857 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V1E2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP6V1E2 - ATPase H+ transporting V1 subunit E2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHOQ - ras homolog family member Q | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012249.3 | 798 | Missense Mutation | AGC,GGC | S,G 54 | NP_036381.2 | |
XM_005264229.2 | 798 | Missense Mutation | AGC,GGC | S,G 54 | XP_005264286.1 | |
XM_011532726.2 | 798 | Missense Mutation | AGC,GGC | S,G 54 | XP_011531028.1 | |
XM_011532728.2 | 798 | Intron | XP_011531030.1 | |||
XM_017003716.1 | 798 | Missense Mutation | AGC,GGC | S,G 54 | XP_016859205.1 |