Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACTCATCAACGATTTCCTTTCCT[C/T]GAAATGAAAATCCCATCCTTAGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608357 | ||||||||||||||||||||
Literature Links: |
SULT1C4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SULT1C4 - sulfotransferase family 1C member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321770.1 | 694 | Silent Mutation | CTC,CTT | L,L 89 | NP_001308699.1 | |
NM_006588.3 | 694 | Silent Mutation | CTC,CTT | L,L 89 | NP_006579.2 | |
XM_017003807.1 | 694 | UTR 5 | XP_016859296.1 |