Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACTGTCCGTCGTCTAATTCCCG[A/G]AGGACGTTGTCACTGCTGCTGAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RFTN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RFTN2 - raftlin family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144629.2 | 2062 | Silent Mutation | CTC,CTT | L,L 481 | NP_653230.2 | |
XM_011510595.2 | 2062 | Silent Mutation | CTC,CTT | L,L 491 | XP_011508897.1 | |
XM_011510596.2 | 2062 | Silent Mutation | CTC,CTT | L,L 490 | XP_011508898.1 | |
XM_011510597.2 | 2062 | Silent Mutation | CTC,CTT | L,L 480 | XP_011508899.1 | |
XM_011510598.2 | 2062 | Silent Mutation | CTC,CTT | L,L 474 | XP_011508900.1 | |
XM_017003332.1 | 2062 | Silent Mutation | CTC,CTT | L,L 464 | XP_016858821.1 |