Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAGCATTGCCCAGTGCACATCCT[C/T]CCAGTGAGTGCGGGCAAGTATGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616978 MIM: 602000 | ||||||||||||||||||||
Literature Links: |
CHCHD5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHCHD5 - coiled-coil-helix-coiled-coil-helix domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304353.1 | 1182 | Missense Mutation | TCC,TTC | S,F 9 | NP_001291282.1 | |
NM_001304354.1 | 1182 | Missense Mutation | TCC,TTC | S,F 9 | NP_001291283.1 | |
NM_032309.3 | 1182 | Missense Mutation | TCC,TTC | S,F 47 | NP_115685.1 | |
XM_005263824.3 | 1182 | Missense Mutation | TCC,TTC | S,F 47 | XP_005263881.1 |
POLR1B - RNA polymerase I subunit B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |