Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCACTCCGGCCCCCTCGAGAAG[A/G]GGTGCCACTGCCTGGAGGGCCCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFT2D3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SFT2D3 - SFT2 domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR33 - WD repeat domain 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006622.2 | 4106 | Intron | NP_001006623.1 | |||
NM_001006623.2 | 4106 | Intron | NP_001006624.1 | |||
NM_018383.4 | 4106 | Missense Mutation | CCT,TCT | P,S 1303 | NP_060853.3 | |
XM_005263697.2 | 4106 | UTR 3 | XP_005263754.1 | |||
XM_011511436.1 | 4106 | Missense Mutation | CCT,TCT | P,S 1303 | XP_011509738.1 | |
XM_017004435.1 | 4106 | Intron | XP_016859924.1 | |||
XM_017004436.1 | 4106 | Intron | XP_016859925.1 |