Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGACGGGGAGCTGAACGTGGACA[G/T]CCTCATCACCCGGCTGCTGGAGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600590 | ||||||||||||||||||||
Literature Links: |
PPP1CB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP1CB - protein phosphatase 1 catalytic subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002709.2 | 189 | Missense Mutation | AGC,ATC | S,I 10 | NP_002700.1 | |
NM_206876.1 | 189 | Missense Mutation | AGC,ATC | S,I 10 | NP_996759.1 |