Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATTTGCAGATAATTTTATATGT[C/G]AAGGAGGAGATGAAAAAAAACAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BZW1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BZW1 - basic leucine zipper and W2 domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207067.1 | 1289 | Silent Mutation | GTC,GTG | V,V 278 | NP_001193996.1 | |
NM_001207069.1 | 1289 | Intron | NP_001193998.1 | |||
NM_001321688.1 | 1289 | Intron | NP_001308617.1 | |||
NM_001321690.1 | 1289 | Intron | NP_001308619.1 | |||
NM_001321691.1 | 1289 | Intron | NP_001308620.1 | |||
NM_001321693.1 | 1289 | Intron | NP_001308622.1 | |||
NM_001321694.1 | 1289 | Intron | NP_001308623.1 | |||
NM_014670.3 | 1289 | Silent Mutation | GTC,GTG | V,V 278 | NP_055485.2 | |
XM_017005393.1 | 1289 | Intron | XP_016860882.1 |
LOC101927795 - uncharacterized LOC101927795 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |