Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCATTATTATTTCAGATTCAAATT[C/T]GGTCTTCATAGATTGGGTTTTAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC173 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC173 - coiled-coil domain containing 173 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHOSPHO2 - phosphatase, orphan 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008489.3 | 604 | Missense Mutation | TCG,TTG | S,L 102 | NP_001008489.1 | |
NM_001199285.1 | 604 | Missense Mutation | TCG,TTG | S,L 102 | NP_001186214.1 | |
NM_001199286.1 | 604 | Missense Mutation | TCG,TTG | S,L 102 | NP_001186215.1 | |
NM_001199287.1 | 604 | Missense Mutation | TCG,TTG | S,L 102 | NP_001186216.1 | |
NM_001199288.1 | 604 | Missense Mutation | TCG,TTG | S,L 102 | NP_001186217.1 |
PHOSPHO2-KLHL23 - PHOSPHO2-KLHL23 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199290.1 | 604 | Intron | NP_001186219.1 |