Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTGCCAAATGTTTGGTGATGAA[A/G]GCAGAAATGAATGGCTCAAAACTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 185535 | ||||||||||||||||||||
Literature Links: |
EPCAM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPCAM - epithelial cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002354.2 | 568 | Silent Mutation | AAA,AAG | K,K 70 | NP_002345.2 |
MIR559 - microRNA 559 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |