Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGACTATGTGCAAATTCCTTATGA[C/G]GTCACTCTCTGGATACTTCTAGCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600531 | ||||||||||||||||||||
Literature Links: |
SLC9A4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC9A4 - solute carrier family 9 member A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011552.3 | 694 | Missense Mutation | GAC,GAG | D,E 72 | NP_001011552.2 | |
XM_011511158.1 | 694 | Missense Mutation | GAC,GAG | D,E 72 | XP_011509460.1 |