Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACAAAGGAGACTGAAATCAATA[C/T]GAGCAAGCCAAAAGCTTCAGGATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604653 | ||||||||||||||||||||
Literature Links: |
SLC40A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC40A1 - solute carrier family 40 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014585.5 | 2056 | Missense Mutation | ATA,GTA | I,V 524 | NP_055400.1 | |
XM_005246505.1 | 2056 | Missense Mutation | ATA,GTA | I,V 484 | XP_005246562.1 | |
XM_017003938.1 | 2056 | Missense Mutation | ATA,GTA | I,V 484 | XP_016859427.1 |