Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAACAACTGAATGTTGTTTAGACA[C/T]GCTAAAGATTGTAAAGAAAAGACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611208 | ||||||||||||||||||||
Literature Links: |
HNRNPLL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HNRNPLL - heterogeneous nuclear ribonucleoprotein L like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142650.1 | 1551 | Missense Mutation | ATG,GTG | M,V 401 | NP_001136122.1 | |
NM_138394.3 | 1551 | Missense Mutation | ATG,GTG | M,V 406 | NP_612403.2 | |
XM_005264639.3 | 1551 | Missense Mutation | ATG,GTG | M,V 406 | XP_005264696.2 | |
XM_005264640.3 | 1551 | Missense Mutation | ATG,GTG | M,V 401 | XP_005264697.2 | |
XM_011533165.2 | 1551 | Missense Mutation | ATG,GTG | M,V 382 | XP_011531467.1 | |
XM_011533166.2 | 1551 | Missense Mutation | ATG,GTG | M,V 406 | XP_011531468.1 |