Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCGAGTTTTTCTTTAACGTGTAC[A/G]TAAACAAAGAGAAAAGACTTCAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142984 MIM: 142986 MIM: 142988 | ||||||||||||||||||||
Literature Links: |
HOXD10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HOXD10 - homeobox D10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXD11 - homeobox D11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021192.2 | 871 | Missense Mutation | ATA,GTA | I,V 291 | NP_067015.2 |
HOXD12 - homeobox D12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107985805 - sine oculis-binding protein homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |