Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCCCGCCAAGACCATCGTGGTGT[A/G]CCGCAACGGGGACCCGTTCTACGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612396 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALLC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALLC - allantoicase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DCDC2C - doublecortin domain containing 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287444.1 | 216 | Missense Mutation | TAC,TGC | Y,C 21 | NP_001274373.1 | |
XM_006711894.3 | 216 | Missense Mutation | TAC,TGC | Y,C 21 | XP_006711957.1 | |
XM_011510393.2 | 216 | Missense Mutation | TAC,TGC | Y,C 21 | XP_011508695.1 | |
XM_011510394.2 | 216 | Missense Mutation | TAC,TGC | Y,C 21 | XP_011508696.1 | |
XM_017004836.1 | 216 | Missense Mutation | TAC,TGC | Y,C 21 | XP_016860325.1 | |
XM_017004837.1 | 216 | Missense Mutation | TAC,TGC | Y,C 21 | XP_016860326.1 | |
XM_017004838.1 | 216 | Missense Mutation | TAC,TGC | Y,C 21 | XP_016860327.1 |