Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGCTCTTTGCTATCCCAGGCAC[C/T]GACAGGGTGGTACGTTTGTTTATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616547 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYG2 - lysozyme g2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175735.3 | 522 | Missense Mutation | AGT,GGT | S,G 137 | NP_783862.2 | |
XM_017003751.1 | 522 | Missense Mutation | AGT,GGT | S,G 137 | XP_016859240.1 |