Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCTCCTCCACATGCTCATCTCC[C/T]CCAGCAGCCTGGGTGGCTGGGAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GPR148 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR148 - G protein-coupled receptor 148 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207364.2 | 404 | Missense Mutation | CCC,TCC | P,S 108 | NP_997247.2 |
POTEI - POTE ankyrin domain family member I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277406.1 | 404 | Intron | NP_001264335.1 | |||
XM_011511670.2 | 404 | Intron | XP_011509972.1 | |||
XM_017004732.1 | 404 | Intron | XP_016860221.1 | |||
XM_017004733.1 | 404 | Intron | XP_016860222.1 | |||
XM_017004734.1 | 404 | Intron | XP_016860223.1 | |||
XM_017004735.1 | 404 | Intron | XP_016860224.1 |