Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTAGAGTTTCCAGACCATCTAT[A/T]ACCCTGAAAGAGAAACAACATAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601951 MIM: 615811 | ||||||||||||||||||||
Literature Links: |
CLK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLK1 - CDC like kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPIL3 - peptidylprolyl isomerase like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032472.3 | 626 | Silent Mutation | GTA,GTT | V,V 125 | NP_115861.1 | |
NM_130906.2 | 626 | Silent Mutation | GTA,GTT | V,V 121 | NP_570981.1 | |
XM_005246651.3 | 626 | Silent Mutation | GTA,GTT | V,V 125 | XP_005246708.1 | |
XM_005246652.4 | 626 | Silent Mutation | GTA,GTT | V,V 121 | XP_005246709.1 | |
XM_011511358.2 | 626 | Silent Mutation | GTA,GTT | V,V 121 | XP_011509660.1 | |
XM_017004352.1 | 626 | Silent Mutation | GTA,GTT | V,V 125 | XP_016859841.1 | |
XM_017004353.1 | 626 | Silent Mutation | GTA,GTT | V,V 125 | XP_016859842.1 | |
XM_017004354.1 | 626 | Intron | XP_016859843.1 | |||
XM_017004355.1 | 626 | Intron | XP_016859844.1 | |||
XM_017004356.1 | 626 | Intron | XP_016859845.1 | |||
XM_017004357.1 | 626 | Silent Mutation | GTA,GTT | V,V 72 | XP_016859846.1 | |
XM_017004358.1 | 626 | Silent Mutation | GTA,GTT | V,V 72 | XP_016859847.1 | |
XM_017004359.1 | 626 | Silent Mutation | GTA,GTT | V,V 72 | XP_016859848.1 | |
XM_017004360.1 | 626 | Silent Mutation | GTA,GTT | V,V 72 | XP_016859849.1 |