Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGTAAAAAGACTACTTACTTACC[C/T]CATGCGCTAACATTTTATAGAGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608960 | ||||||||||||||||||||
Literature Links: |
ARL5A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARL5A - ADP ribosylation factor like GTPase 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037174.1 | 401 | Missense Mutation | GAG,GGG | E,G 76 | NP_001032251.1 | |
NM_012097.3 | 401 | Missense Mutation | GAG,GGG | E,G 113 | NP_036229.1 | |
NM_177985.2 | 401 | Missense Mutation | GAG,GGG | E,G 76 | NP_817114.2 |