Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCACTTCCTAGAGTTGCTTCTG[C/T]ATGTTTTAAGAGTTCATACTGATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609281 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MOB1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MOB1A - MOB kinase activator 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317110.1 | 384 | Missense Mutation | ACA,GCA | T,A 31 | NP_001304039.1 | |
NM_001317111.1 | 384 | Missense Mutation | ACA,GCA | T,A 74 | NP_001304040.1 | |
NM_001317112.1 | 384 | Missense Mutation | ACA,GCA | T,A 32 | NP_001304041.1 | |
NM_018221.4 | 384 | Missense Mutation | ACA,GCA | T,A 32 | NP_060691.2 | |
XM_011532948.2 | 384 | Missense Mutation | ACA,GCA | T,A 74 | XP_011531250.1 |