Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAGCCGGGCGGCGCGGCCCTGCA[A/G]GGGCTCAGACCGCCGCCGCCGCCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 150390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100271832 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100271832 - uncharacterized LOC100271832 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LTBP1 - latent transforming growth factor beta binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000627.3 | 484 | Intron | NP_000618.3 | |||
NM_001166264.1 | 484 | Intron | NP_001159736.1 | |||
NM_001166265.1 | 484 | Intron | NP_001159737.1 | |||
NM_001166266.1 | 484 | Intron | NP_001159738.1 | |||
NM_206943.2 | 484 | Silent Mutation | CAA,CAG | Q,Q 96 | NP_996826.2 | |
XM_005264317.3 | 484 | Silent Mutation | CAA,CAG | Q,Q 96 | XP_005264374.1 | |
XM_005264318.3 | 484 | Silent Mutation | CAA,CAG | Q,Q 96 | XP_005264375.1 | |
XM_011532853.2 | 484 | Silent Mutation | CAA,CAG | Q,Q 96 | XP_011531155.1 | |
XM_011532855.2 | 484 | Silent Mutation | CAA,CAG | Q,Q 96 | XP_011531157.1 | |
XM_011532856.2 | 484 | Silent Mutation | CAA,CAG | Q,Q 96 | XP_011531158.1 | |
XM_011532857.2 | 484 | Silent Mutation | CAA,CAG | Q,Q 96 | XP_011531159.1 | |
XM_011532858.2 | 484 | Silent Mutation | CAA,CAG | Q,Q 96 | XP_011531160.1 | |
XM_011532859.2 | 484 | Silent Mutation | CAA,CAG | Q,Q 96 | XP_011531161.1 | |
XM_011532860.2 | 484 | Silent Mutation | CAA,CAG | Q,Q 96 | XP_011531162.1 | |
XM_011532861.2 | 484 | Silent Mutation | CAA,CAG | Q,Q 96 | XP_011531163.1 | |
XM_011532862.2 | 484 | Silent Mutation | CAA,CAG | Q,Q 96 | XP_011531164.1 | |
XM_017004108.1 | 484 | Silent Mutation | CAA,CAG | Q,Q 96 | XP_016859597.1 | |
XM_017004109.1 | 484 | Silent Mutation | CAA,CAG | Q,Q 96 | XP_016859598.1 | |
XM_017004110.1 | 484 | Silent Mutation | CAA,CAG | Q,Q 96 | XP_016859599.1 |