Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCCCATCACTGTGGCCGAGTAC[A/G]TGAAGGAGGTGTTGACTAATCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612828 MIM: 615898 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEBPZ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEBPZ - CCAAT/enhancer binding protein zeta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005760.2 | 262 | Intron | NP_005751.2 |
NDUFAF7 - NADH:ubiquinone oxidoreductase complex assembly factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083946.1 | 262 | Missense Mutation | ATG,GTG | M,V 63 | NP_001077415.1 | |
NM_144736.4 | 262 | Missense Mutation | ATG,GTG | M,V 63 | NP_653337.1 | |
XM_005264410.1 | 262 | Missense Mutation | ATG,GTG | M,V 21 | XP_005264467.1 | |
XM_005264411.1 | 262 | Missense Mutation | ATG,GTG | M,V 63 | XP_005264468.1 | |
XM_011532956.1 | 262 | UTR 5 | XP_011531258.1 |