Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGAGGGACCAGGACAGGAGACC[A/G]GGATGAGAAGCTAACAAGAGGCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610767 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG16L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATG16L1 - autophagy related 16 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190266.1 | Intron | NP_001177195.1 | ||||
NM_001190267.1 | Intron | NP_001177196.1 | ||||
NM_017974.3 | Intron | NP_060444.3 | ||||
NM_030803.6 | Intron | NP_110430.5 | ||||
NM_198890.2 | Intron | NP_942593.2 | ||||
XM_005246082.2 | Intron | XP_005246139.1 | ||||
XM_005246084.2 | Intron | XP_005246141.1 | ||||
XM_005246086.2 | Intron | XP_005246143.1 | ||||
XM_006712608.2 | Intron | XP_006712671.1 |