Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGCAGTGTTTGAATGGATCTGA[C/T]AGCGACTTAATAGGGTTAGAGGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP36 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP36 - cilia and flagella associated protein 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282761.1 | Intron | NP_001269690.1 | ||||
NM_080667.6 | Intron | NP_542398.3 | ||||
XM_011532499.1 | Intron | XP_011530801.1 | ||||
XM_017003254.1 | Intron | XP_016858743.1 | ||||
XM_017003255.1 | Intron | XP_016858744.1 |