Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTCTGATTCCTTCATTAATGTCC[A/G]GTAAGTAAATGTGCACACCCAGTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GPATCH11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPATCH11 - G-patch domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278505.1 | 192 | Missense Mutation | AGG,GGG | R,G 3 | NP_001265434.1 | |
NM_001322249.1 | 192 | Missense Mutation | CAA,CGA | Q,R 42 | NP_001309178.1 | |
NM_174931.3 | 192 | Missense Mutation | CAA,CGA | Q,R 42 | NP_777591.3 | |
XM_017003748.1 | 192 | Intron | XP_016859237.1 |
HEATR5B - HEAT repeat containing 5B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |