Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGTAGAAGTAGTGGTAGAAATT[G/T]TACAGTGGGCTGCAAGAGAAACCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605538 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NTSR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NTSR2 - neurotensin receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012344.3 | 1073 | Nonsense Mutation | TAA,TAC | *,Y 333 | NP_036476.1 | |
XM_005246156.3 | 1073 | Missense Mutation | AAA,ACA | K,T 242 | XP_005246213.1 | |
XM_006711876.3 | 1073 | Missense Mutation | AAA,ACA | K,T 314 | XP_006711939.1 | |
XM_006711877.3 | 1073 | Missense Mutation | AAA,ACA | K,T 311 | XP_006711940.1 | |
XM_017003738.1 | 1073 | Missense Mutation | AAA,ACA | K,T 331 | XP_016859227.1 |