Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCTGAAGCAGTTTCATCCTCCTC[C/G]TCCTCTGGATTATATCCACCAGTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608381 MIM: 192975 | ||||||||||||||||||||
Literature Links: |
CERKL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CERKL - ceramide kinase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030311.2 | 1499 | Missense Mutation | GAC,GAG | D,E 510 | NP_001025482.1 | |
NM_001030312.2 | 1499 | Missense Mutation | GAC,GAG | D,E 371 | NP_001025483.1 | |
NM_001030313.2 | 1499 | Missense Mutation | GAC,GAG | D,E 415 | NP_001025484.1 | |
NM_001160277.1 | 1499 | Missense Mutation | GAC,GAG | D,E 466 | NP_001153749.1 | |
NM_201548.4 | 1499 | Missense Mutation | GAC,GAG | D,E 484 | NP_963842.1 |
ITGA4 - integrin subunit alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000885.5 | 1499 | Intron | NP_000876.3 | |||
NM_001316312.1 | 1499 | Intron | NP_001303241.1 |