Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTCACTTACCATCTGTGGATCA[C/T]TGGATCGGCTCACCCAACCAGATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602559 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
XPO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
XPO1 - exportin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003400.3 | 3028 | Missense Mutation | AAT,AGT | N,S 767 | NP_003391.1 | |
XM_005264544.1 | 3028 | Missense Mutation | AAT,AGT | N,S 752 | XP_005264601.1 | |
XM_005264546.1 | 3028 | Missense Mutation | AAT,AGT | N,S 636 | XP_005264603.1 | |
XM_006712094.2 | 3028 | Missense Mutation | AAT,AGT | N,S 767 | XP_006712157.1 | |
XM_011533097.1 | 3028 | Missense Mutation | AAT,AGT | N,S 767 | XP_011531399.1 | |
XM_011533098.1 | 3028 | Missense Mutation | AAT,AGT | N,S 722 | XP_011531400.1 | |
XM_011533099.2 | 3028 | Missense Mutation | AAT,AGT | N,S 701 | XP_011531401.1 |