Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAAATCCAGCTTGGTATTTAAA[A/G]CAGAAAATAACATAGTTTTAGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607030 MIM: 608169 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GCA - grancalcin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNH7 - potassium voltage-gated channel subfamily H member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033272.3 | Intron | NP_150375.2 | ||||
NM_173162.2 | Intron | NP_775185.1 | ||||
XM_011512109.2 | Intron | XP_011510411.1 | ||||
XM_017005218.1 | Intron | XP_016860707.1 | ||||
XM_017005219.1 | Intron | XP_016860708.1 | ||||
XM_017005220.1 | Intron | XP_016860709.1 | ||||
XM_017005221.1 | Intron | XP_016860710.1 |