Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCAAAACCTGAAGGGGCCCTCCA[C/G]AACAATGATGGGCTTTATGATCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 185535 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPCAM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPCAM - epithelial cell adhesion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002354.2 | 625 | Silent Mutation | CAC,CAG | H,Q 89 | NP_002345.2 |
MIR559 - microRNA 559 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |