Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCAAAGTCCTCTCGAAGCGGCG[C/G]TGTCTCCGCACGGAGAGATCGGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EVA1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EVA1A - eva-1 homolog A, regulator of programmed cell death | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135032.1 | 778 | Silent Mutation | CAC,CAG | H,Q 100 | NP_001128504.1 | |
NM_032181.2 | 778 | Silent Mutation | CAC,CAG | H,Q 100 | NP_115557.1 |