Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAAATACCTTCTGCAACAACTCA[C/T]TTCTAGAAACAGACATCATGGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606997 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1D - C1D nuclear receptor corepressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190263.1 | Intron | NP_001177192.1 | ||||
NM_001190265.1 | Intron | NP_001177194.1 | ||||
NM_006333.3 | Intron | NP_006324.1 | ||||
NM_173177.2 | Intron | NP_775269.1 |