Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGGACGGCAGCCCAGGGGGCGG[C/T]GGGCTGAGGAGCAGTGCGGGGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604994 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SIX2 - SIX homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016932.4 | 998 | Silent Mutation | CCA,CCG | P,P 235 | NP_058628.3 | |
XM_005264100.3 | 998 | Silent Mutation | CCA,CCG | P,P 237 | XP_005264157.1 |