Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATAGGCTGTGCCATCCAGGGCC[A/G]TGGCCTCCTTGTCTCTGCTGTCCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604024 | ||||||||||||||||||||
Literature Links: |
SLC5A6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC5A6 - solute carrier family 5 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021095.2 | 2131 | Missense Mutation | ACG,ATG | T,M 614 | NP_066918.2 | |
XM_006712128.1 | 2131 | Missense Mutation | ACG,ATG | T,M 614 | XP_006712191.1 | |
XM_006712129.1 | 2131 | Missense Mutation | ACG,ATG | T,M 614 | XP_006712192.1 | |
XM_006712130.1 | 2131 | Missense Mutation | ACG,ATG | T,M 614 | XP_006712193.1 | |
XM_011533146.2 | 2131 | Missense Mutation | ACG,ATG | T,M 391 | XP_011531448.1 | |
XM_017005216.1 | 2131 | Missense Mutation | ACG,ATG | T,M 405 | XP_016860705.1 |