Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGCTACTGGGGTTAGCTACCTT[G/T]TTGGAGCTGGCAAAGCCATTGGTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600291 MIM: 611504 | ||||||||||||||||||||
Literature Links: |
ADCY3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADCY3 - adenylate cyclase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320613.1 | 3314 | Missense Mutation | AAA,AAC | K,N 1001 | NP_001307542.1 | |
NM_004036.4 | 3314 | Missense Mutation | AAA,AAC | K,N 1000 | NP_004027.2 | |
XM_006711925.1 | 3314 | Missense Mutation | AAA,AAC | K,N 1023 | XP_006711988.1 | |
XM_011532489.1 | 3314 | Missense Mutation | AAA,AAC | K,N 1042 | XP_011530791.1 | |
XM_011532490.1 | 3314 | Missense Mutation | AAA,AAC | K,N 1041 | XP_011530792.1 | |
XM_011532491.1 | 3314 | Missense Mutation | AAA,AAC | K,N 1020 | XP_011530793.1 | |
XM_011532492.2 | 3314 | Missense Mutation | AAA,AAC | K,N 1042 | XP_011530794.1 | |
XM_011532493.1 | 3314 | Missense Mutation | AAA,AAC | K,N 996 | XP_011530795.1 | |
XM_011532494.1 | 3314 | Missense Mutation | AAA,AAC | K,N 976 | XP_011530796.1 | |
XM_011532496.1 | 3314 | Missense Mutation | AAA,AAC | K,N 801 | XP_011530798.1 | |
XM_017003186.1 | 3314 | Missense Mutation | AAA,AAC | K,N 1022 | XP_016858675.1 | |
XM_017003187.1 | 3314 | Missense Mutation | AAA,AAC | K,N 1019 | XP_016858676.1 | |
XM_017003188.1 | 3314 | Missense Mutation | AAA,AAC | K,N 1041 | XP_016858677.1 | |
XM_017003189.1 | 3314 | Missense Mutation | AAA,AAC | K,N 995 | XP_016858678.1 | |
XM_017003190.1 | 3314 | Missense Mutation | AAA,AAC | K,N 954 | XP_016858679.1 | |
XM_017003191.1 | 3314 | Missense Mutation | AAA,AAC | K,N 830 | XP_016858680.1 | |
XM_017003192.1 | 3314 | Missense Mutation | AAA,AAC | K,N 760 | XP_016858681.1 | |
XM_017003193.1 | 3314 | Missense Mutation | AAA,AAC | K,N 759 | XP_016858682.1 |
CENPO - centromere protein O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199803.2 | 3314 | Intron | NP_001186732.1 | |||
NM_001322101.1 | 3314 | Intron | NP_001309030.1 | |||
NM_024322.3 | 3314 | Intron | NP_077298.1 |