Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCCTTTCTCTCCTCCTAAAAGA[C/T]GAGCTCATTCCTGGCCCCCCTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611367 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNPEP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNPEP - aspartyl aminopeptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319116.1 | Intron | NP_001306045.1 | ||||
NM_001319117.1 | Intron | NP_001306046.1 | ||||
NM_001319118.1 | Intron | NP_001306047.1 | ||||
NM_001319119.1 | Intron | NP_001306048.1 | ||||
NM_001319120.1 | Intron | NP_001306049.1 | ||||
NM_001319121.1 | Intron | NP_001306050.1 | ||||
NM_001319122.1 | Intron | NP_001306051.1 | ||||
NM_012100.3 | Intron | NP_036232.2 |