Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCCGAGTGCCTCACCTCGAGGAC[A/C]ACTTTGCGCATGCGCCCCAGCTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611504 | ||||||||||||||||||||
Literature Links: |
CENPO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CENPO - centromere protein O | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199803.2 | 248 | UTR 5 | NP_001186732.1 | |||
NM_001322101.1 | 248 | UTR 5 | NP_001309030.1 | |||
NM_024322.3 | 248 | UTR 5 | NP_077298.1 |
PTRHD1 - peptidyl-tRNA hydrolase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013663.1 | 248 | Silent Mutation | NP_001013685.1 |