Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACATTGAGTTCTAGAACTATCCC[A/G]AGGCAATCAATCAGCAATGACACAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616526 | ||||||||||||||||||||
Literature Links: |
SLC38A11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC38A11 - solute carrier family 38 member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199148.1 | 1551 | Silent Mutation | CTC,CTT | L,L 302 | NP_001186077.1 | |
NM_173512.2 | 1551 | Silent Mutation | CTC,CTT | L,L 280 | NP_775783.1 | |
XM_005246350.4 | 1551 | Silent Mutation | CTC,CTT | L,L 358 | XP_005246407.1 | |
XM_006712337.3 | 1551 | Silent Mutation | CTC,CTT | L,L 304 | XP_006712400.1 | |
XM_011510737.2 | 1551 | Silent Mutation | CTC,CTT | L,L 336 | XP_011509039.1 | |
XM_011510742.2 | 1551 | Silent Mutation | CTC,CTT | L,L 163 | XP_011509044.1 | |
XM_017003455.1 | 1551 | Silent Mutation | CTC,CTT | L,L 384 | XP_016858944.1 | |
XM_017003456.1 | 1551 | Silent Mutation | CTC,CTT | L,L 362 | XP_016858945.1 | |
XM_017003457.1 | 1551 | Silent Mutation | CTC,CTT | L,L 359 | XP_016858946.1 | |
XM_017003458.1 | 1551 | Silent Mutation | CTC,CTT | L,L 328 | XP_016858947.1 | |
XM_017003459.1 | 1551 | Silent Mutation | CTC,CTT | L,L 306 | XP_016858948.1 | |
XM_017003460.1 | 1551 | Silent Mutation | CTC,CTT | L,L 384 | XP_016858949.1 | |
XM_017003461.1 | 1551 | Silent Mutation | CTC,CTT | L,L 384 | XP_016858950.1 | |
XM_017003462.1 | 1551 | Silent Mutation | CTC,CTT | L,L 384 | XP_016858951.1 | |
XM_017003463.1 | 1551 | Silent Mutation | CTC,CTT | L,L 236 | XP_016858952.1 | |
XM_017003464.1 | 1551 | Silent Mutation | CTC,CTT | L,L 189 | XP_016858953.1 | |
XM_017003465.1 | 1551 | Silent Mutation | CTC,CTT | L,L 189 | XP_016858954.1 |