Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGAAGCGGCTGGACATGAGGCTA[C/T]AGAGCACGGGGTTGGCCGCCGAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604153 | ||||||||||||||||||||
Literature Links: |
NMUR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NMUR1 - neuromedin U receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006056.4 | 1165 | Missense Mutation | TAT,TGT | Y,C 356 | NP_006047.3 | |
XM_006712195.3 | 1165 | Intron | XP_006712258.1 | |||
XM_006712196.3 | 1165 | Intron | XP_006712259.1 | |||
XM_011510487.2 | 1165 | Missense Mutation | TAT,TGT | Y,C 333 | XP_011508789.1 | |
XM_011510488.2 | 1165 | Intron | XP_011508790.1 | |||
XM_011510489.2 | 1165 | UTR 3 | XP_011508791.1 |