Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTAGTCCACATCGCTCTCCAGCG[A/G]GAAGCTCTCCTCCTTGCCCTTCGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601135 | ||||||||||||||||||||
Literature Links: |
GBX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GBX2 - gastrulation brain homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301687.1 | 636 | Missense Mutation | CCC,CTC | P,L 212 | NP_001288616.1 | |
NM_001485.3 | 636 | Missense Mutation | CCG,TCG | P,S 200 | NP_001476.2 |
LOC105373944 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |