Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCACTGAGATCTTTCACCACTCC[C/T]TCTTTGACCAGCTTTAAGAGGAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604402 | ||||||||||||||||||||
Literature Links: |
ST3GAL5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ST3GAL5 - ST3 beta-galactoside alpha-2,3-sialyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042437.1 | 1455 | Silent Mutation | GAA,GAG | E,E 381 | NP_001035902.1 | |
NM_003896.3 | 1455 | Silent Mutation | GAA,GAG | E,E 404 | NP_003887.3 | |
XM_005264630.4 | 1455 | Silent Mutation | GAA,GAG | E,E 351 | XP_005264687.1 | |
XM_011533143.2 | 1455 | Silent Mutation | GAA,GAG | E,E 276 | XP_011531445.1 | |
XM_017005199.1 | 1455 | Silent Mutation | GAA,GAG | E,E 376 | XP_016860688.1 | |
XM_017005200.1 | 1455 | Silent Mutation | GAA,GAG | E,E 376 | XP_016860689.1 | |
XM_017005201.1 | 1455 | Silent Mutation | GAA,GAG | E,E 376 | XP_016860690.1 | |
XM_017005202.1 | 1455 | Silent Mutation | GAA,GAG | E,E 323 | XP_016860691.1 | |
XM_017005203.1 | 1455 | Silent Mutation | GAA,GAG | E,E 276 | XP_016860692.1 | |
XM_017005204.1 | 1455 | Silent Mutation | GAA,GAG | E,E 276 | XP_016860693.1 | |
XM_017005205.1 | 1455 | Silent Mutation | GAA,GAG | E,E 276 | XP_016860694.1 | |
XM_017005206.1 | 1455 | Silent Mutation | GAA,GAG | E,E 276 | XP_016860695.1 | |
XM_017005207.1 | 1455 | Silent Mutation | GAA,GAG | E,E 276 | XP_016860696.1 | |
XM_017005208.1 | 1455 | Silent Mutation | GAA,GAG | E,E 276 | XP_016860697.1 | |
XM_017005209.1 | 1455 | Silent Mutation | GAA,GAG | E,E 276 | XP_016860698.1 | |
XM_017005210.1 | 1455 | Silent Mutation | GAA,GAG | E,E 276 | XP_016860699.1 | |
XM_017005211.1 | 1455 | Silent Mutation | GAA,GAG | E,E 276 | XP_016860700.1 | |
XM_017005212.1 | 1455 | Silent Mutation | GAA,GAG | E,E 276 | XP_016860701.1 | |
XM_017005213.1 | 1455 | Silent Mutation | GAA,GAG | E,E 276 | XP_016860702.1 | |
XM_017005214.1 | 1455 | Silent Mutation | GAA,GAG | E,E 223 | XP_016860703.1 | |
XM_017005215.1 | 1455 | Silent Mutation | GAA,GAG | E,E 223 | XP_016860704.1 |