Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTCTTGAACCGCTAAAAGCTCT[A/G]TGTTACTGGCTGGTCTGTTTCTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEATR5B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HEATR5B - HEAT repeat containing 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019024.2 | 6406 | Missense Mutation | ACA,ATA | T,I 1947 | NP_061897.1 | |
XM_006712034.2 | 6406 | Missense Mutation | ACA,ATA | T,I 1946 | XP_006712097.1 | |
XM_006712035.3 | 6406 | Missense Mutation | ACA,ATA | T,I 1935 | XP_006712098.1 | |
XM_011532934.2 | 6406 | Missense Mutation | ACA,ATA | T,I 1947 | XP_011531236.1 | |
XM_011532935.2 | 6406 | Missense Mutation | ACA,ATA | T,I 1330 | XP_011531237.1 | |
XM_017004378.1 | 6406 | Missense Mutation | ACA,ATA | T,I 1858 | XP_016859867.1 | |
XM_017004379.1 | 6406 | Missense Mutation | ACA,ATA | T,I 1822 | XP_016859868.1 | |
XM_017004380.1 | 6406 | Missense Mutation | ACA,ATA | T,I 1822 | XP_016859869.1 |