Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGTTATCAGTTGGCCTACTTTC[C/T]AAAACCACGACCCTGAAAGTAATAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604921 | ||||||||||||||||||||
Literature Links: |
CDKL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDKL4 - cyclin dependent kinase like 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAP4K3 - mitogen-activated protein kinase kinase kinase kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270425.1 | 2935 | Silent Mutation | TTA,TTG | L,L 849 | NP_001257354.1 | |
NM_003618.3 | 2935 | Silent Mutation | TTA,TTG | L,L 870 | NP_003609.2 |