Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGGAAGAGTTCCTTCACGTTATT[A/G]CCTGTGTGCTGAGCTCGAGCTGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607633 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
XDH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
XDH - xanthine dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000379.3 | 3943 | Silent Mutation | GGC,GGT | G,G 1288 | NP_000370.2 | |
XM_011533095.2 | 3943 | Silent Mutation | GGC,GGT | G,G 1287 | XP_011531397.1 | |
XM_011533096.2 | 3943 | Intron | XP_011531398.1 |