Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTTGCAAGATATGAGAAACATG[A/G]AGGTTTTCTGACCCTGCTTCCAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 177010 | ||||||||||||||||||||
Literature Links: |
SERPINE2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SERPINE2 - serpin family E member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136528.1 | 1060 | Silent Mutation | CTC,CTT | L,L 333 | NP_001130000.1 | |
NM_001136530.1 | 1060 | Silent Mutation | CTC,CTT | L,L 345 | NP_001130002.1 | |
NM_006216.3 | 1060 | Silent Mutation | CTC,CTT | L,L 334 | NP_006207.1 | |
XM_005246641.2 | 1060 | Silent Mutation | CTC,CTT | L,L 346 | XP_005246698.1 | |
XM_017004329.1 | 1060 | Silent Mutation | CTC,CTT | L,L 334 | XP_016859818.1 | |
XM_017004330.1 | 1060 | Silent Mutation | CTC,CTT | L,L 334 | XP_016859819.1 | |
XM_017004331.1 | 1060 | Silent Mutation | CTC,CTT | L,L 333 | XP_016859820.1 | |
XM_017004332.1 | 1060 | Silent Mutation | CTC,CTT | L,L 333 | XP_016859821.1 |