Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCTTTAATGGAATATTCTAACG[C/T]AGTACATAGTTATGATATAGTACGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R21 - protein phosphatase 1 regulatory subunit 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135629.2 | Intron | NP_001129101.1 | ||||
NM_001193475.1 | Intron | NP_001180404.1 | ||||
NM_152994.4 | Intron | NP_694539.1 | ||||
XM_017003301.1 | Intron | XP_016858790.1 | ||||
XM_017003302.1 | Intron | XP_016858791.1 |