Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCGCGCGAAGCGGCGGCCGCACA[C/G]GTCGCAAGCAAAAGGCTTCTCGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 128992 MIM: 609108 | ||||||||||||||||||||
Literature Links: |
EGR4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EGR4 - early growth response 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001965.3 | 1708 | Missense Mutation | CTG,GTG | L,V 545 | NP_001956.3 |
FBXO41 - F-box protein 41 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |