Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGACCTGTTCTGAGCCCAGGAT[A/C]ATCAAGGCAGGCTCAGGTTCCAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614765 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STRN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STRN - striatin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003162.3 | 11603 | UTR 3 | NP_003153.2 | |||
XM_005264519.4 | 11603 | UTR 3 | XP_005264576.1 | |||
XM_011533073.2 | 11603 | UTR 3 | XP_011531375.1 |