Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAAGTGATACGTGACTAATTTAC[A/G]TGAAATTTATACATTCTTTATCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612246 MIM: 613334 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD302 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD302 - CD302 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198763.1 | 6674 | UTR 3 | NP_001185692.1 | |||
NM_001198764.1 | 6674 | UTR 3 | NP_001185693.1 | |||
NM_014880.4 | 6674 | UTR 3 | NP_055695.2 |
LY75-CD302 - LY75-CD302 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198759.1 | 6674 | UTR 3 | NP_001185688.1 | |||
NM_001198760.1 | 6674 | UTR 3 | NP_001185689.1 |
MARCH7 - membrane associated ring-CH-type finger 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |