Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGATATACTAAGAAGAAAACAGA[A/G]GCTTGCCTTTTACAATTAGTGGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609778 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
XIRP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
XIRP2 - xin actin binding repeat containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079810.3 | Intron | NP_001073278.1 | ||||
NM_001199143.1 | Intron | NP_001186072.1 | ||||
NM_001199144.1 | Intron | NP_001186073.1 | ||||
NM_001199145.1 | Intron | NP_001186074.1 | ||||
NM_152381.5 | Intron | NP_689594.4 | ||||
XM_017003309.1 | Intron | XP_016858798.1 |