Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAACTCAGTCCTGTTGGAATCAG[C/T]ATTTTATTATATGAAGTTGTTATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDKL4 - cyclin dependent kinase like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009565.1 | 1241 | UTR 3 | NP_001009565.1 | |||
XM_011532815.2 | 1241 | Intron | XP_011531117.1 | |||
XM_011532816.2 | 1241 | Intron | XP_011531118.1 | |||
XM_011532817.2 | 1241 | Intron | XP_011531119.1 | |||
XM_011532819.2 | 1241 | Intron | XP_011531121.1 | |||
XM_017003981.1 | 1241 | Intron | XP_016859470.1 | |||
XM_017003982.1 | 1241 | Intron | XP_016859471.1 | |||
XM_017003983.1 | 1241 | Intron | XP_016859472.1 | |||
XM_017003984.1 | 1241 | Intron | XP_016859473.1 | |||
XM_017003985.1 | 1241 | Intron | XP_016859474.1 |